Product Details

SNP ID
rs188185160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:104114633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGAAAGATGCAAACCAGACTCG[C/G]TTGCAACAGCTTCTTTTAGGCGCTG
Phenotype
MIM: 300941
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC286437 PubMed Links

Gene Details

Gene
LOC286437
Gene Name
uncharacterized LOC286437
There are no transcripts associated with this gene.

Gene
SLC25A53
Gene Name
solute carrier family 25 member 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012755.4 1728 Intron NP_001012773.2
XM_005262129.4 1728 Intron XP_005262186.1
XM_011530952.2 1728 Intron XP_011529254.1
XM_011530953.2 1728 Intron XP_011529255.1
XM_017029514.1 1728 Intron XP_016885003.1
XM_017029515.1 1728 Intron XP_016885004.1
XM_017029516.1 1728 Intron XP_016885005.1
Gene
ZCCHC18
Gene Name
zinc finger CCHC-type containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143978.2 1728 Silent Mutation CGC,CGG R174R NP_001137450.1
XM_011531012.2 1728 Silent Mutation CGC,CGG R174R XP_011529314.1

View Full Product Details