Product Details

SNP ID
rs188227976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:45907218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGGCTGAGCCCAGCCCGGGGGT[A/G]CAGAGCCCCCAGCATGGCGAGAACC
Phenotype
MIM: 604641 MIM: 603360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf94 PubMed Links

Gene Details

Gene
C11orf94
Gene Name
chromosome 11 open reading frame 94
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080446.2 65 Missense Mutation NP_001073915.2
Gene
MAPK8IP1
Gene Name
mitogen-activated protein kinase 8 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005456.3 65 Intron NP_005447.1
XM_005253226.3 65 Intron XP_005253283.1
Gene
PEX16
Gene Name
peroxisomal biogenesis factor 16
There are no transcripts associated with this gene.

View Full Product Details