Product Details

SNP ID
rs188225608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:151180186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCATCCACTTCGTCCTCCCTCT[C/T]CTCATCGTGGGTTTCTGCTACGTGA
Phenotype
MIM: 300207
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR50 PubMed Links

Gene Details

Gene
GPR50
Gene Name
G protein-coupled receptor 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004224.3 741 Silent Mutation CTC,CTT L201L NP_004215.2
XM_011531216.2 741 Intron XP_011529518.1
Gene
GPR50-AS1
Gene Name
GPR50 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details