Product Details

SNP ID
rs187958429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:39012710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATACCCGGCTTGTGCAACAAGAG[C/T]GGAGTTGAAATTGGAACTGAATGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PROSER1 PubMed Links

Gene Details

Gene
PROSER1
Gene Name
proline and serine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025138.4 3349 Missense Mutation NP_079414.3
XM_005266545.3 3349 Missense Mutation XP_005266602.1
XM_011535239.2 3349 Missense Mutation XP_011533541.1

View Full Product Details