Product Details
- SNP ID
-
rs187977939
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:127048535 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCTGGGCCCCGCCGTCATGGGAC[C/T]CTCTCAGTGAAGTTCTCGGGGAAGA
- Phenotype
-
MIM: 601248
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BIN1
PubMed Links
Gene Details
- Gene
- BIN1
- Gene Name
- bridging integrator 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001320632.1 |
2275 |
Silent Mutation |
AGA,AGG |
R422R |
NP_001307561.1 |
NM_001320633.1 |
2275 |
Silent Mutation |
AGA,AGG |
R468R |
NP_001307562.1 |
NM_001320634.1 |
2275 |
Silent Mutation |
AGA,AGG |
R383R |
NP_001307563.1 |
NM_001320640.1 |
2275 |
Silent Mutation |
AGA,AGG |
R511R |
NP_001307569.1 |
NM_001320641.1 |
2275 |
Silent Mutation |
AGA,AGG |
R560R |
NP_001307570.1 |
NM_001320642.1 |
2275 |
Silent Mutation |
AGA,AGG |
R564R |
NP_001307571.1 |
NM_004305.3 |
2275 |
Silent Mutation |
AGA,AGG |
R452R |
NP_004296.1 |
NM_139343.2 |
2275 |
Silent Mutation |
AGA,AGG |
R591R |
NP_647593.1 |
NM_139344.2 |
2275 |
Silent Mutation |
AGA,AGG |
R548R |
NP_647594.1 |
NM_139345.2 |
2275 |
Silent Mutation |
AGA,AGG |
R504R |
NP_647595.1 |
NM_139346.2 |
2275 |
Silent Mutation |
AGA,AGG |
R495R |
NP_647596.1 |
NM_139347.2 |
2275 |
Silent Mutation |
AGA,AGG |
R516R |
NP_647597.1 |
NM_139348.2 |
2275 |
Silent Mutation |
AGA,AGG |
R480R |
NP_647598.1 |
NM_139349.2 |
2275 |
Silent Mutation |
AGA,AGG |
R473R |
NP_647599.1 |
NM_139350.2 |
2275 |
Silent Mutation |
AGA,AGG |
R437R |
NP_647600.1 |
NM_139351.2 |
2275 |
Silent Mutation |
AGA,AGG |
R407R |
NP_647601.1 |
XM_005263642.3 |
2275 |
Silent Mutation |
AGA,AGG |
R606R |
XP_005263699.1 |
XM_005263643.3 |
2275 |
Silent Mutation |
AGA,AGG |
R575R |
XP_005263700.1 |
XM_005263645.2 |
2275 |
Silent Mutation |
AGA,AGG |
R551R |
XP_005263702.1 |
XM_006712425.3 |
2275 |
Silent Mutation |
AGA,AGG |
R562R |
XP_006712488.1 |
XM_006712426.3 |
2275 |
Silent Mutation |
AGA,AGG |
R562R |
XP_006712489.1 |
XM_006712427.2 |
2275 |
Silent Mutation |
AGA,AGG |
R536R |
XP_006712490.1 |
XM_006712428.3 |
2275 |
Silent Mutation |
AGA,AGG |
R526R |
XP_006712491.1 |
XM_006712429.3 |
2275 |
Silent Mutation |
AGA,AGG |
R519R |
XP_006712492.1 |
XM_006712431.3 |
2275 |
Silent Mutation |
AGA,AGG |
R496R |
XP_006712494.1 |
XM_006712432.3 |
2275 |
Silent Mutation |
AGA,AGG |
R483R |
XP_006712495.1 |
XM_006712433.3 |
2275 |
Silent Mutation |
AGA,AGG |
R453R |
XP_006712496.1 |
XM_006712434.3 |
2275 |
Silent Mutation |
AGA,AGG |
R438R |
XP_006712497.1 |
XM_011510975.2 |
2275 |
Silent Mutation |
AGA,AGG |
R579R |
XP_011509277.1 |
XM_017003819.1 |
2275 |
Silent Mutation |
AGA,AGG |
R582R |
XP_016859308.1 |
XM_017003820.1 |
2275 |
Silent Mutation |
AGA,AGG |
R516R |
XP_016859309.1 |
XM_017003821.1 |
2275 |
Silent Mutation |
AGA,AGG |
R481R |
XP_016859310.1 |
XM_017003822.1 |
2275 |
Silent Mutation |
AGA,AGG |
R450R |
XP_016859311.1 |
XM_017003823.1 |
2275 |
Silent Mutation |
AGA,AGG |
R449R |
XP_016859312.1 |
XM_017003824.1 |
2275 |
Silent Mutation |
AGA,AGG |
R426R |
XP_016859313.1 |
XM_017003825.1 |
2275 |
Silent Mutation |
AGA,AGG |
R414R |
XP_016859314.1 |
XM_017003826.1 |
2275 |
Silent Mutation |
AGA,AGG |
R413R |
XP_016859315.1 |
XM_017003827.1 |
2275 |
Silent Mutation |
AGA,AGG |
R382R |
XP_016859316.1 |
XM_017003828.1 |
2275 |
Silent Mutation |
AGA,AGG |
R367R |
XP_016859317.1 |
View Full Product Details