Product Details
- SNP ID
-
rs187973880
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:86532226 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACTCACACGGGCGTCCCCGGGGA[C/G]GTTCCCAACGTAAACATCGGCTGCA
- Phenotype
-
MIM: 616820
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MTHFSD
PubMed Links
Gene Details
- Gene
- MTHFSD
- Gene Name
- methenyltetrahydrofolate synthetase domain containing
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001159377.1 |
988 |
Missense Mutation |
CTC,GTC |
L313V |
NP_001152849.1 |
NM_001159378.1 |
988 |
Missense Mutation |
CTC,GTC |
L313V |
NP_001152850.1 |
NM_001159379.1 |
988 |
Missense Mutation |
CTC,GTC |
L312V |
NP_001152851.1 |
NM_001159380.1 |
988 |
Missense Mutation |
CTC,GTC |
L293V |
NP_001152852.1 |
NM_022764.2 |
988 |
Missense Mutation |
CTC,GTC |
L312V |
NP_073601.2 |
XM_005256101.1 |
988 |
Missense Mutation |
CTC,GTC |
L293V |
XP_005256158.1 |
XM_005256105.1 |
988 |
Missense Mutation |
CTC,GTC |
L171V |
XP_005256162.1 |
XM_005256106.1 |
988 |
Missense Mutation |
CTC,GTC |
L171V |
XP_005256163.1 |
XM_006721247.3 |
988 |
Missense Mutation |
CTC,GTC |
L150V |
XP_006721310.1 |
XM_011523280.2 |
988 |
Missense Mutation |
CTC,GTC |
L293V |
XP_011521582.1 |
XM_011523282.2 |
988 |
Intron |
|
|
XP_011521584.1 |
XM_011523283.2 |
988 |
Intron |
|
|
XP_011521585.1 |
XM_011523284.2 |
988 |
Intron |
|
|
XP_011521586.1 |
XM_011523285.1 |
988 |
Missense Mutation |
CTC,GTC |
L150V |
XP_011521587.1 |
XM_011523286.1 |
988 |
Missense Mutation |
CTC,GTC |
L150V |
XP_011521588.1 |
XM_011523287.1 |
988 |
Missense Mutation |
CTC,GTC |
L150V |
XP_011521589.1 |
XM_011523288.1 |
988 |
Missense Mutation |
CTC,GTC |
L150V |
XP_011521590.1 |
XM_011523289.1 |
988 |
Missense Mutation |
CTC,GTC |
L150V |
XP_011521591.1 |
XM_017023571.1 |
988 |
Missense Mutation |
CTC,GTC |
L275V |
XP_016879060.1 |
XM_017023572.1 |
988 |
Missense Mutation |
CTC,GTC |
L274V |
XP_016879061.1 |
XM_017023573.1 |
988 |
Missense Mutation |
CTC,GTC |
L171V |
XP_016879062.1 |
XM_017023574.1 |
988 |
Missense Mutation |
CTC,GTC |
L150V |
XP_016879063.1 |
View Full Product Details