Product Details
- SNP ID
-
rs188107887
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:4907531 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTTATTATAAAGACAGAGCCCTC[A/G]CTTCATGAGCCCATGTATTATTTCC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR51A7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7108225] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR51A7
- Gene Name
- olfactory receptor family 51 subfamily A member 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004749.1 |
162 |
Silent Mutation |
TCA,TCG |
S54S |
NP_001004749.1 |
- Gene
- OR51G2
- Gene Name
- olfactory receptor family 51 subfamily G member 2
There are no transcripts associated with this gene.
View Full Product Details