Product Details

SNP ID
rs188099318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120766371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACGCCACAGTAGCAGTGAGCAG[C/G]CCTGTGAGGAGAGAGAGGCATCTGT
Phenotype
MIM: 608240
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPPL3 PubMed Links

Gene Details

Gene
SPPL3
Gene Name
signal peptide peptidase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139015.4 1460 Silent Mutation GGC,GGG G325G NP_620584.2
XM_011537925.2 1460 Silent Mutation GGC,GGG G325G XP_011536227.1

View Full Product Details