Product Details
- SNP ID
-
rs187831058
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:143569711 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTCTCACACCTTCCTGCGATTCA[G/T]CGCGAGGTGGCCCAGGCCCAGCAGG
- Phenotype
-
MIM: 130592
MIM: 611552
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EEF1D
PubMed Links
Gene Details
- Gene
- EEF1D
- Gene Name
- eukaryotic translation elongation factor 1 delta
There are no transcripts associated with this gene.
- Gene
- GSDMD
- Gene Name
- gasdermin D
There are no transcripts associated with this gene.
- Gene
- MROH6
- Gene Name
- maestro heat like repeat family member 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001100878.1 |
1326 |
Missense Mutation |
ATG,CTG |
M430L |
NP_001094348.1 |
XM_006716615.2 |
1326 |
Missense Mutation |
ATG,CTG |
M430L |
XP_006716678.1 |
XM_011517214.1 |
1326 |
Missense Mutation |
ATG,CTG |
M430L |
XP_011515516.1 |
XM_011517215.1 |
1326 |
Missense Mutation |
ATG,CTG |
M430L |
XP_011515517.1 |
XM_011517216.2 |
1326 |
Missense Mutation |
ATG,CTG |
M430L |
XP_011515518.1 |
XM_011517217.1 |
1326 |
Missense Mutation |
ATG,CTG |
M430L |
XP_011515519.1 |
XM_011517218.1 |
1326 |
Missense Mutation |
ATG,CTG |
M430L |
XP_011515520.1 |
XM_011517219.1 |
1326 |
Missense Mutation |
ATG,CTG |
M430L |
XP_011515521.1 |
XM_011517220.1 |
1326 |
Missense Mutation |
ATG,CTG |
M430L |
XP_011515522.1 |
XM_011517221.2 |
1326 |
Missense Mutation |
ATG,CTG |
M430L |
XP_011515523.1 |
XM_011517222.1 |
1326 |
Missense Mutation |
ATG,CTG |
M430L |
XP_011515524.1 |
XM_011517223.1 |
1326 |
Missense Mutation |
ATG,CTG |
M430L |
XP_011515525.1 |
- Gene
- NAPRT
- Gene Name
- nicotinate phosphoribosyltransferase
There are no transcripts associated with this gene.
View Full Product Details