Product Details

SNP ID
rs187870139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:119557536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAGAAAGTTAAATCTCGCTCTC[A/G]TATGTATGCAGGTATGTCTTGAGTG
Phenotype
MIM: 601060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENPP2 PubMed Links

Gene Details

Gene
ENPP2
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040092.2 2663 Silent Mutation NP_001035181.1
NM_001130863.2 2663 Silent Mutation NP_001124335.1
NM_006209.4 2663 Silent Mutation NP_006200.3
XM_006716584.1 2663 Nonsense Mutation XP_006716647.1
XM_006716585.1 2663 Nonsense Mutation XP_006716648.1
XM_006716587.1 2663 Nonsense Mutation XP_006716650.1
XM_017013571.1 2663 Nonsense Mutation XP_016869060.1
XM_017013572.1 2663 Silent Mutation XP_016869061.1
XM_017013573.1 2663 Nonsense Mutation XP_016869062.1
XM_017013574.1 2663 Nonsense Mutation XP_016869063.1
XM_017013575.1 2663 Nonsense Mutation XP_016869064.1

View Full Product Details