Product Details
- SNP ID
-
rs188479152
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:126396023 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGATGTCTGAGATACCGTCCACTA[C/T]AGTCTCCAAGAACATGACCAATGAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC37-AS1
PubMed Links
Gene Details
- Gene
- CCDC37-AS1
- Gene Name
- CCDC37 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
- Gene
- CFAP100
- Gene Name
- cilia and flagella associated protein 100
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182628.2 |
273 |
Missense Mutation |
ACA,ATA |
T8I |
NP_872434.2 |
XM_006713623.2 |
273 |
Missense Mutation |
ACA,ATA |
T8I |
XP_006713686.1 |
XM_017006320.1 |
273 |
Missense Mutation |
ACA,ATA |
T8I |
XP_016861809.1 |
XM_017006321.1 |
273 |
Missense Mutation |
ACA,ATA |
T8I |
XP_016861810.1 |
XM_017006322.1 |
273 |
Missense Mutation |
ACA,ATA |
T8I |
XP_016861811.1 |
XM_017006323.1 |
273 |
Missense Mutation |
ACA,ATA |
T8I |
XP_016861812.1 |
XM_017006324.1 |
273 |
Missense Mutation |
ACA,ATA |
T8I |
XP_016861813.1 |
XM_017006325.1 |
273 |
Missense Mutation |
ACA,ATA |
T8I |
XP_016861814.1 |
XM_017006326.1 |
273 |
Missense Mutation |
ACA,ATA |
T8I |
XP_016861815.1 |
XM_017006327.1 |
273 |
Missense Mutation |
ACA,ATA |
T8I |
XP_016861816.1 |
View Full Product Details