Product Details

SNP ID
rs188463327
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:173186843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTCCTTTTGGGAAGTGAGGAT[A/G]AAACCTTTCTCCTTCTTATATTCTA
Phenotype
MIM: 603594
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFSF4 PubMed Links

Gene Details

Gene
TNFSF4
Gene Name
tumor necrosis factor superfamily member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297562.1 849 Silent Mutation TTC,TTT F25F NP_001284491.1
NM_003326.4 849 Silent Mutation TTC,TTT F75F NP_003317.1
XM_011509964.2 849 Silent Mutation TTC,TTT F171F XP_011508266.2
XM_017002228.1 849 Silent Mutation TTC,TTT F107F XP_016857717.1
XM_017002229.1 849 Silent Mutation TTC,TTT F86F XP_016857718.1
XM_017002230.1 849 Silent Mutation TTC,TTT F84F XP_016857719.1

View Full Product Details