Product Details

SNP ID
rs188425418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:34024017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTACCTGGTGTCAATTGGAGTGA[G/T]CAAACCTGCCATTTGGATCTTTGGA
Phenotype
MIM: 603313
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALG10 PubMed Links

Gene Details

Gene
ALG10
Gene Name
ALG10, alpha-1,2-glucosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032834.3 Intron NP_116223.3

View Full Product Details