Product Details

SNP ID
rs188702994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:44977772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGCTGCTGGCCACTGAAACGTG[C/T]GGTGATGTCTCGCAGGGACACAGCA
Phenotype
MIM: 611309 MIM: 610559
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf10 PubMed Links

Gene Details

Gene
C10orf10
Gene Name
chromosome 10 open reading frame 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007021.3 549 Missense Mutation ACA,GCA T87A NP_008952.1
Gene
RASSF4
Gene Name
Ras association domain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032023.3 549 Intron NP_114412.2
XM_005271833.4 549 Intron XP_005271890.1
XM_005271835.4 549 Intron XP_005271892.1
XM_006718013.3 549 Intron XP_006718076.1
XM_006718014.3 549 Intron XP_006718077.1
XM_011540255.2 549 Intron XP_011538557.1
XM_017016754.1 549 Intron XP_016872243.1
XM_017016755.1 549 Intron XP_016872244.1
XM_017016756.1 549 Intron XP_016872245.1
XM_017016757.1 549 Intron XP_016872246.1
XM_017016758.1 549 Intron XP_016872247.1

View Full Product Details