Product Details

SNP ID
rs188741039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36748063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGATGTCGAATTAGTTCTGAGC[C/T]GCAAGTAAAAGATTTCCCACATTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF850 PubMed Links

Gene Details

Gene
ZNF850
Gene Name
zinc finger protein 850
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193552.1 3040 Missense Mutation AGC,GGC S993G NP_001180481.1
NM_001267779.1 3040 Missense Mutation AGC,GGC S961G NP_001254708.1

View Full Product Details