Product Details

SNP ID
rs188690808
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:15872927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTGCACAGAAAGCTCACAACTC[A/G]GTCAACAAAATGGGTGACAGAGACC
Phenotype
MIM: 613484
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPEN PubMed Links

Gene Details

Gene
SPEN
Gene Name
spen family transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015001.2 399 Silent Mutation TCA,TCG S65S NP_055816.2

View Full Product Details