Product Details

SNP ID
rs188843575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130067230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGTGGCGCCGGGTTGGTCGGAC[A/G]GTTGGCCCCAGCCAGAAGTTGGGGG
Phenotype
MIM: 300688 MIM: 300775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCORL1 PubMed Links

Gene Details

Gene
BCORL1
Gene Name
BCL6 corepressor-like 1
There are no transcripts associated with this gene.

Gene
ELF4
Gene Name
E74 like ETS transcription factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127197.1 2204 Missense Mutation CGT,TGT R495C NP_001120669.1
NM_001421.3 2204 Missense Mutation CGT,TGT R495C NP_001412.1
XM_005262389.3 2204 Missense Mutation CGT,TGT R495C XP_005262446.1
XM_011531307.2 2204 Missense Mutation CGT,TGT R427C XP_011529609.1
XM_011531308.2 2204 Missense Mutation CGT,TGT R418C XP_011529610.1

View Full Product Details