Product Details

SNP ID
rs188799901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66703319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTGGCGAAGTCGTGTGGTCCCC[A/G]ACCGGGAGGAAGGCGGCAGCGGCGG
Phenotype
MIM: 602931
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMAD6 PubMed Links

Gene Details

Gene
SMAD6
Gene Name
SMAD family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005585.4 984 Missense Mutation AAC,GAC N21D NP_005576.3
XM_011521561.2 984 Intron XP_011519863.1

View Full Product Details