Product Details
- SNP ID
-
rs188506223
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:152154746 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTCTTCATGAGTTTGCCTGTCTC[A/G]TCTCTGATGGTTCTGCTCGTCTTCA
- Phenotype
-
MIM: 613259
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RPTN
PubMed Links
Gene Details
- Gene
- RPTN
- Gene Name
- repetin
There are no transcripts associated with this gene.
View Full Product Details