Product Details

SNP ID
rs188545155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:80263725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCATGTCTCCAAGGAGGAAACCC[C/T]CAAGTTCTGCAGCCAGTGCGGAGAG
Phenotype
MIM: 613768
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF213 PubMed Links

Gene Details

Gene
RNF213
Gene Name
ring finger protein 213
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256071.2 235 Missense Mutation CCC,CTC P15L NP_001243000.2
NM_020954.3 235 Missense Mutation CCC,CTC P15L NP_066005.2
XM_005257545.4 235 Missense Mutation CCC,CTC P15L XP_005257602.2
XM_005257546.4 235 Missense Mutation CCC,CTC P15L XP_005257603.2
XM_006721995.3 235 Missense Mutation CCC,CTC P15L XP_006722058.1
XM_011525084.2 235 Missense Mutation CCC,CTC P15L XP_011523386.1
XM_011525086.2 235 Missense Mutation CCC,CTC P15L XP_011523388.1
XM_011525087.2 235 Missense Mutation CCC,CTC P15L XP_011523389.1
XM_017024905.1 235 Intron XP_016880394.1

View Full Product Details