Product Details

SNP ID
rs188572043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39265546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGATTCAGATGAGGAGAAATCAA[C/T]GTGGACTGAGCGCCCACACAGGACA
Phenotype
MIM: 601470
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CX3CR1 PubMed Links

Gene Details

Gene
CX3CR1
Gene Name
C-X3-C motif chemokine receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171171.1 1056 Missense Mutation ATT,GTT I322V NP_001164642.1
NM_001171172.1 1056 Missense Mutation ATT,GTT I322V NP_001164643.1
NM_001171174.1 1056 Missense Mutation ATT,GTT I354V NP_001164645.1
NM_001337.3 1056 Missense Mutation ATT,GTT I322V NP_001328.1

View Full Product Details