Product Details

SNP ID
rs188632527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28715042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCTCCAACTCACCAGTGAGAGA[A/T]GAGACTGCCCAGTACTCAGCCTTCA
Phenotype
MIM: 610917 MIM: 602326
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NARR PubMed Links

Gene Details

Gene
NARR
Gene Name
nine-amino acid residue-repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256281.1 766 Intron NP_001243210.1
Gene
PROCA1
Gene Name
protein interacting with cyclin A1
There are no transcripts associated with this gene.

Gene
RAB34
Gene Name
RAB34, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142624.2 766 Silent Mutation TCA,TCT S247S NP_001136096.2
NM_001142625.2 766 Intron NP_001136097.2
NM_001144942.1 766 Silent Mutation TCA,TCT S190S NP_001138414.1
NM_001144943.1 766 Silent Mutation TCA,TCT S255S NP_001138415.1
NM_001256276.1 766 Silent Mutation TCA,TCT S176S NP_001243205.1
NM_001256277.1 766 Silent Mutation TCA,TCT S198S NP_001243206.1
NM_001256278.1 766 Silent Mutation TCA,TCT S198S NP_001243207.1
NM_031934.5 766 Silent Mutation TCA,TCT S198S NP_114140.4
Gene
RPL23A
Gene Name
ribosomal protein L23a
There are no transcripts associated with this gene.

Gene
SNORD42A
Gene Name
small nucleolar RNA, C/D box 42A
There are no transcripts associated with this gene.

Gene
SNORD42B
Gene Name
small nucleolar RNA, C/D box 42B
There are no transcripts associated with this gene.

Gene
SNORD4A
Gene Name
small nucleolar RNA, C/D box 4A
There are no transcripts associated with this gene.

Gene
SNORD4B
Gene Name
small nucleolar RNA, C/D box 4B
There are no transcripts associated with this gene.

Gene
TLCD1
Gene Name
TLC domain containing 1
There are no transcripts associated with this gene.

View Full Product Details