Product Details

SNP ID
rs188652308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4716566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCACCCGCCTGCAGGACCGGCT[C/G]CTGAGGAAGCTGGGCCAGCATGCCC
Phenotype
MIM: 107941
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARRB2 PubMed Links

Gene Details

Gene
ARRB2
Gene Name
arrestin beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257328.1 543 Silent Mutation CTC,CTG L105L NP_001244257.1
NM_001257329.1 543 Silent Mutation CTC,CTG L105L NP_001244258.1
NM_001257330.1 543 Silent Mutation CTC,CTG L105L NP_001244259.1
NM_001257331.1 543 Silent Mutation CTC,CTG L90L NP_001244260.1
NM_004313.3 543 Silent Mutation CTC,CTG L105L NP_004304.1
NM_199004.1 543 Silent Mutation CTC,CTG L90L NP_945355.1
XM_006721520.1 543 UTR 5 XP_006721583.1
XM_011523858.2 543 Silent Mutation CTC,CTG L136L XP_011522160.1
XM_017024645.1 543 UTR 5 XP_016880134.1

View Full Product Details