Product Details

SNP ID
rs188412721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:131504284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCAACTTGAGCCTTGTGGCCCT[G/T]ACTGGGATGGGGTTACTGAGCCGGC
Phenotype
MIM: 607423
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
POMT1 PubMed Links

Gene Details

Gene
POMT1
Gene Name
protein O-mannosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077365.1 268 Silent Mutation CTG,CTT L22L NP_001070833.1
NM_001077366.1 268 Intron NP_001070834.1
NM_001136113.1 268 Silent Mutation CTG,CTT L22L NP_001129585.1
NM_001136114.1 268 Intron NP_001129586.1
NM_007171.3 268 Silent Mutation CTG,CTT L22L NP_009102.3
XM_005272156.1 268 Silent Mutation CTG,CTT L22L XP_005272213.1
XM_005272158.1 268 Intron XP_005272215.1
XM_005272159.1 268 Intron XP_005272216.1
XM_005272162.2 268 UTR 5 XP_005272219.1
XM_006716932.1 268 UTR 5 XP_006716995.1
XM_011518140.1 268 Silent Mutation CTG,CTT L22L XP_011516442.1
XM_011518141.1 268 Silent Mutation CTG,CTT L22L XP_011516443.1
XM_011518142.1 268 Intron XP_011516444.1
XM_011518143.1 268 Silent Mutation CTG,CTT L22L XP_011516445.1
XM_011518145.1 268 Intron XP_011516447.1
XM_017014203.1 268 UTR 5 XP_016869692.1
XM_017014204.1 268 Intron XP_016869693.1
XM_017014205.1 268 UTR 5 XP_016869694.1
Gene
PRRC2B
Gene Name
proline rich coiled-coil 2B
There are no transcripts associated with this gene.

View Full Product Details