Product Details

SNP ID
rs189006292
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:27613040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGCCAGGTCTGTTCCATGGCCTC[A/G]TACAGGGATGCTGGGCAGCCTGGAG
Phenotype
MIM: 615790 MIM: 164940
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AHDC1 PubMed Links

Gene Details

Gene
AHDC1
Gene Name
AT-hook DNA binding motif containing 1
There are no transcripts associated with this gene.

Gene
FGR
Gene Name
FGR proto-oncogene, Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042729.1 1692 Silent Mutation TAC,TAT Y488Y NP_001036194.1
NM_001042747.1 1692 Silent Mutation TAC,TAT Y488Y NP_001036212.1
NM_005248.2 1692 Silent Mutation TAC,TAT Y488Y NP_005239.1
XM_006710452.2 1692 Silent Mutation TAC,TAT Y488Y XP_006710515.1
XM_011541010.1 1692 Silent Mutation TAC,TAT Y488Y XP_011539312.1
XM_011541011.2 1692 Silent Mutation TAC,TAT Y419Y XP_011539313.1
XM_011541012.1 1692 UTR 3 XP_011539314.1
XM_011541013.2 1692 Intron XP_011539315.1
XM_011541014.2 1692 Silent Mutation TAC,TAT Y284Y XP_011539316.1
XM_017000673.1 1692 Silent Mutation TAC,TAT Y284Y XP_016856162.1
XM_017000674.1 1692 Silent Mutation TAC,TAT Y284Y XP_016856163.1

View Full Product Details