Product Details

SNP ID
rs188968729
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140668669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATGTTCTGGCCACTGGGGATGA[C/G]ACAGGTGGTATCTGTCTCTGGGACC
Phenotype
MIM: 609385 MIM: 142810 MIM: 600549
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
DND1 PubMed Links
Additional Information
For this assay, SNP(s) [rs2251860] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DND1
Gene Name
DND microRNA-mediated repression inhibitor 1
There are no transcripts associated with this gene.

Gene
HARS
Gene Name
histidyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
IK
Gene Name
IK cytokine, down-regulator of HLA II
There are no transcripts associated with this gene.

Gene
WDR55
Gene Name
WD repeat domain 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017706.4 552 Missense Mutation GAC,GAG D146E NP_060176.2
XM_005268469.2 552 Missense Mutation GAC,GAG D146E XP_005268526.1
XM_017009600.1 552 UTR 5 XP_016865089.1

View Full Product Details