Product Details

SNP ID
rs189005809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7593180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTGTAGACACATCTGAGCTGGG[C/G]CCTGAAGAACACAATGGGATTTATT
Phenotype
MIM: 605339 MIM: 604041 MIM: 601297
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FXR2 PubMed Links

Gene Details

Gene
FXR2
Gene Name
FMR1 autosomal homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004860.3 1697 Silent Mutation GGC,GGG G444G NP_004851.2
Gene
LOC100996842
Gene Name
uncharacterized LOC100996842
There are no transcripts associated with this gene.

Gene
MPDU1
Gene Name
mannose-P-dolichol utilization defect 1
There are no transcripts associated with this gene.

Gene
SOX15
Gene Name
SRY-box 15
There are no transcripts associated with this gene.

View Full Product Details