Product Details

SNP ID
rs189013609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102729737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGATTTATGTTTGCTTTGGAAA[C/G]AATCCAATCTTTCTGACTCATAATC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC168 PubMed Links

Gene Details

Gene
CCDC168
Gene Name
coiled-coil domain containing 168
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146197.1 21100 Missense Mutation TCT,TGT S6987C NP_001139669.1
XM_011521106.1 21100 Missense Mutation TCT,TGT S6947C XP_011519408.1

View Full Product Details