Product Details

SNP ID
rs189260891
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:50419514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGCGCTGCCTGGGCAGCTGCC[A/G]CCTGCCTCTTTGGAGGCCCGAGCTA
Phenotype
MIM: 614804
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DMRTA2 PubMed Links

Gene Details

Gene
DMRTA2
Gene Name
DMRT like family A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032110.2 1909 Silent Mutation NP_115486.1
XM_011541937.2 1909 Silent Mutation XP_011540239.1

View Full Product Details