Product Details

SNP ID
rs189403310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6620892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCGGGTGCACGATGGCGGCGTA[A/G]CGGTCCACGTTGATGAGCATCAGGA
Phenotype
MIM: 606926
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LPAR5 PubMed Links

Gene Details

Gene
LPAR5
Gene Name
lysophosphatidic acid receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142961.1 638 Silent Mutation CGC,CGT R119R NP_001136433.1
NM_020400.5 638 Silent Mutation CGC,CGT R119R NP_065133.1

View Full Product Details