Product Details

SNP ID
rs189385964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122910854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCTCCCACCTTGAATTCCATGG[A/G]TGTGTACTTTTCATTCTTCGGGGTG
Phenotype
MIM: 609298
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEMA5B PubMed Links

Gene Details

Gene
SEMA5B
Gene Name
semaphorin 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031702.3 3720 Missense Mutation NP_001026872.2
NM_001256346.1 3720 Missense Mutation NP_001243275.1
NM_001256347.1 3720 Missense Mutation NP_001243276.1
NM_001256348.1 3720 Missense Mutation NP_001243277.1
XM_017006638.1 3720 Missense Mutation XP_016862127.1
XM_017006639.1 3720 Intron XP_016862128.1

View Full Product Details