Product Details

SNP ID
rs189427123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37171479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGACAGCACCGTGTGTGTCCTGAG[A/C]CTTCGCACGCATGGCTTGGAAGGTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SULT6B1 PubMed Links

Gene Details

Gene
SULT6B1
Gene Name
sulfotransferase family 6B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032377.1 643 Missense Mutation GCT,TCT A208S NP_001027549.1

View Full Product Details