Product Details

SNP ID
rs189316749
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20892002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCATCGGAGTAGATTCCGGGTG[A/C]CTTTACTCCACTGTGACCTCATAAA
Phenotype
MIM: 131398
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
RNASE3 PubMed Links

Gene Details

Gene
RNASE3
Gene Name
ribonuclease A family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002935.2 370 Missense Mutation ACT,CCT T106P NP_002926.2

View Full Product Details