Product Details

SNP ID
rs189368533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132748415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGGCTTCAGAACACCTTTTACTT[A/C]TCCCAGTCCACTTTTAACCTGGCCC
Phenotype
MIM: 608418
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCNI2 PubMed Links

Gene Details

Gene
CCNI2
Gene Name
cyclin I family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039780.3 595 Silent Mutation TTA,TTC L166F NP_001034869.1
NM_001287252.1 595 Silent Mutation TTA,TTC L166F NP_001274181.1
NM_001287253.1 595 Silent Mutation TTA,TTC L167F NP_001274182.1
XM_017009725.1 595 Missense Mutation TTA,TTC L167F XP_016865214.1
Gene
SEPT8
Gene Name
septin 8
There are no transcripts associated with this gene.

View Full Product Details