Product Details
- SNP ID
-
rs189330558
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:72139786 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAGGTGGCCAAGTTCTATGCTGC[G/T]GCCATGAATGCTGGGGCCCCGTCTC
- Phenotype
-
MIM: 602469
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FOLR3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34970007] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FOLR3
- Gene Name
- folate receptor 3
View Full Product Details