Product Details
- SNP ID
-
rs189051470
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:17638829 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTCACCGCCGCTGGGGGACCCTGT[C/T]GGAAGCAACTGCCGCCGCCGCCTCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BCL2L13
PubMed Links
Gene Details
- Gene
- BCL2L13
- Gene Name
- BCL2 like 13
View Full Product Details