Product Details

SNP ID
rs189093134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53076980 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATGCTGTTCTGCATCAGCGTCA[C/T]CCGCTACATGGCCATCGCCCACCAC
Phenotype
MIM: 300253 MIM: 300564
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR173 PubMed Links

Gene Details

Gene
GPR173
Gene Name
G protein-coupled receptor 173
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018969.5 634 Missense Mutation ACC,ATC T120I NP_061842.1
XM_011530798.2 634 Missense Mutation ACC,ATC T120I XP_011529100.1
Gene
TSPYL2
Gene Name
TSPY like 2
There are no transcripts associated with this gene.

View Full Product Details