Product Details

SNP ID
rs189046619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89433143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTAATTCTCTTGCCACAGAATAA[G/T]CCACTGATCCATTGGTCCATAGCTG
Phenotype
MIM: 611910
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC16A12 PubMed Links

Gene Details

Gene
SLC16A12
Gene Name
solute carrier family 16 member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213606.3 1661 Missense Mutation GAT,GCT D491A NP_998771.3
XM_017016237.1 1661 Missense Mutation GAT,GCT D491A XP_016871726.1
XM_017016238.1 1661 Missense Mutation GAT,GCT D491A XP_016871727.1
XM_017016239.1 1661 Missense Mutation GAT,GCT D491A XP_016871728.1

View Full Product Details