Product Details

SNP ID
rs189178959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64251746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGAGTAAGTTCATCAAATGGGAC[C/G]AGGTAAGCGCGCGGGCCCCTGCTCC
Phenotype
MIM: 186946 MIM: 600230 MIM: 601140
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FKBP2 PubMed Links

Gene Details

Gene
FKBP2
Gene Name
FK506 binding protein 2
There are no transcripts associated with this gene.

Gene
LOC105369340
Gene Name
uncharacterized LOC105369340
There are no transcripts associated with this gene.

Gene
PLCB3
Gene Name
phospholipase C beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000932.2 224 Missense Mutation CAG,GAG Q33E NP_000923.1
NM_001184883.1 224 Missense Mutation CAG,GAG Q33E NP_001171812.1
NM_001316314.1 224 Missense Mutation CAG,GAG Q33E NP_001303243.1
XM_011545101.2 224 Missense Mutation CAG,GAG Q33E XP_011543403.1
XM_017017925.1 224 Missense Mutation CAG,GAG Q33E XP_016873414.1
Gene
PPP1R14B
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14B
There are no transcripts associated with this gene.

View Full Product Details