Product Details

SNP ID
rs189169594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40981657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATACTAAGCCCACGTATTGAATGG[A/G]AAGAAAGGTCTGTAAGTGGCTCTCT
Phenotype
MIM: 610169
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INO80 PubMed Links

Gene Details

Gene
INO80
Gene Name
INO80 complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017553.2 Intron NP_060023.1
XM_011521685.2 Intron XP_011519987.1
XM_011521686.2 Intron XP_011519988.1

View Full Product Details