Product Details

SNP ID
rs189219270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184346741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGCACTGTCTCGGAAGCTGGC[A/G]AGGGGCGGCCGGACTTTCACACCCT
Phenotype
MIM: 600570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCN2 PubMed Links

Gene Details

Gene
CLCN2
Gene Name
chloride voltage-gated channel 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171087.2 2683 Silent Mutation CTC,CTT L837L NP_001164558.1
NM_001171088.2 2683 Silent Mutation CTC,CTT L810L NP_001164559.1
NM_001171089.2 2683 Silent Mutation CTC,CTT L825L NP_001164560.1
NM_004366.5 2683 Silent Mutation CTC,CTT L854L NP_004357.3
XM_006713489.1 2683 Silent Mutation CTC,CTT L819L XP_006713552.1
XM_006713490.2 2683 Silent Mutation CTC,CTT L468L XP_006713553.1
XM_011512401.1 2683 Intron XP_011510703.1
Gene
FAM131A
Gene Name
family with sequence similarity 131 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171093.1 2683 Intron NP_001164564.1
NM_144635.4 2683 Intron NP_653236.3
XM_005247113.2 2683 Intron XP_005247170.1
XM_005247114.3 2683 Intron XP_005247171.1
XM_011512413.2 2683 Intron XP_011510715.1

View Full Product Details