Product Details

SNP ID
rs188944676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31304419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGGGCAAGTTAAGTGAGTCAAG[C/T]GGTGGAAATGATTGTATTATCTCAG
Phenotype
MIM: 158381 MIM: 613113 MIM: 164345
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EVI2B PubMed Links

Gene Details

Gene
EVI2B
Gene Name
ecotropic viral integration site 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006495.3 1456 Silent Mutation CCA,CCG P397P NP_006486.3
XM_005257946.3 1456 Silent Mutation CCA,CCG P412P XP_005258003.1
Gene
NF1
Gene Name
neurofibromin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000267.3 1456 Intron NP_000258.1
NM_001042492.2 1456 Intron NP_001035957.1
NM_001128147.2 1456 Intron NP_001121619.1
XM_005257983.2 1456 Intron XP_005258040.1
XM_005257984.2 1456 Intron XP_005258041.1
XM_006721922.2 1456 Intron XP_006721985.1
XM_006721924.2 1456 Intron XP_006721987.1
XM_006721925.2 1456 Intron XP_006721988.1
XM_006721926.3 1456 Intron XP_006721989.1
XM_011524852.2 1456 Intron XP_011523154.1
XM_011524857.2 1456 Intron XP_011523159.1
XM_017024689.1 1456 Intron XP_016880178.1
XM_017024690.1 1456 Intron XP_016880179.1
Gene
OMG
Gene Name
oligodendrocyte myelin glycoprotein
There are no transcripts associated with this gene.

View Full Product Details