Product Details

SNP ID
rs188940530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24440202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGCAGCATGATGGCACGCTGTC[A/G]CCCAAAGACAGCTTGCACCACAGCG
Phenotype
MIM: 616162
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KHNYN PubMed Links

Gene Details

Gene
KHNYN
Gene Name
KH and NYN domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290256.1 755 UTR 3 NP_001277185.1
NM_001290257.1 755 UTR 3 NP_001277186.1
NM_015299.2 755 UTR 3 NP_056114.1
XM_005267474.4 755 UTR 3 XP_005267531.1
XM_011536590.2 755 UTR 3 XP_011534892.1
XM_017021114.1 755 Intron XP_016876603.1
Gene
LOC101927045
Gene Name
uncharacterized LOC101927045
There are no transcripts associated with this gene.

Gene
SDR39U1
Gene Name
short chain dehydrogenase/reductase family 39U member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290292.1 755 Nonsense Mutation CGA,TGA R173* NP_001277221.1
NM_001290293.2 755 Nonsense Mutation CGA,TGA R147* NP_001277222.1
NM_001308075.1 755 Nonsense Mutation CGA,TGA R173* NP_001295004.1
NM_020195.2 755 Nonsense Mutation CGA,TGA R255* NP_064580.2
XM_011536975.1 755 Nonsense Mutation CGA,TGA R278* XP_011535277.1
XM_011536976.1 755 Nonsense Mutation CGA,TGA R213* XP_011535278.1

View Full Product Details