Product Details

SNP ID
rs189550077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17356709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGATCACACCCCTGAATGACAA[C/T]GTCACCATATTCTGCAATATCTTTT
Phenotype
MIM: 613714
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105376575 PubMed Links

Gene Details

Gene
LOC105376575
Gene Name
uncharacterized LOC105376575
There are no transcripts associated with this gene.

Gene
NCR3LG1
Gene Name
natural killer cell cytotoxicity receptor 3 ligand 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202439.2 1943 Silent Mutation AAC,AAT N43N NP_001189368.1
XM_011520072.2 1943 Silent Mutation AAC,AAT N14N XP_011518374.1
XM_011520073.2 1943 Silent Mutation AAC,AAT N14N XP_011518375.1
XM_011520074.2 1943 Silent Mutation AAC,AAT N14N XP_011518376.1
XM_011520075.2 1943 Silent Mutation AAC,AAT N14N XP_011518377.1

View Full Product Details