Product Details

SNP ID
rs189555627
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41970288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGTCGCTTTCGGCAGCCTCGTA[C/T]GCCAGTGAGATGGCAGGGACCTAGG
Phenotype
MIM: 182350
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ATP1A3 PubMed Links

Gene Details

Gene
ATP1A3
Gene Name
ATPase Na+/K+ transporting subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256213.1 2678 Silent Mutation GCA,GCG A824A NP_001243142.1
NM_001256214.1 2678 Silent Mutation GCA,GCG A826A NP_001243143.1
NM_152296.4 2678 Silent Mutation GCA,GCG A813A NP_689509.1

View Full Product Details