Product Details

SNP ID
rs189523393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:87901927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCTTACCTTATGTTTTATTTC[C/T]GGCAAGATGTCCACCAGCCTCTGCA
Phenotype
MIM: 114761
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CA5A PubMed Links

Gene Details

Gene
CA5A
Gene Name
carbonic anhydrase 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001739.1 728 Silent Mutation CCA,CCG P201P NP_001730.1
XM_005256134.4 728 Intron XP_005256191.1
XM_011523309.2 728 Silent Mutation CCA,CCG P201P XP_011521611.1
XM_011523310.2 728 Silent Mutation CCA,CCG P201P XP_011521612.1
XM_017023646.1 728 Silent Mutation CCA,CCG P201P XP_016879135.1

View Full Product Details