Product Details

SNP ID
rs189969139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:171063946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACTTGGCTACTTCCTCCAGATC[G/T]CACGGATGCAAAAAATACCTGTTTG
Phenotype
MIM: 610005
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TNIK PubMed Links

Gene Details

Gene
TNIK
Gene Name
TRAF2 and NCK interacting kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161560.2 4276 Silent Mutation AGA,CGA R1332R NP_001155032.1
NM_001161561.2 4276 Silent Mutation AGA,CGA R1311R NP_001155033.1
NM_001161562.2 4276 Silent Mutation AGA,CGA R1303R NP_001155034.1
NM_001161563.2 4276 Silent Mutation AGA,CGA R1285R NP_001155035.1
NM_001161564.2 4276 Silent Mutation AGA,CGA R1277R NP_001155036.1
NM_001161565.2 4276 Silent Mutation AGA,CGA R1256R NP_001155037.1
NM_001161566.2 4276 Silent Mutation AGA,CGA R1248R NP_001155038.1
NM_015028.3 4276 Silent Mutation AGA,CGA R1340R NP_055843.1

View Full Product Details