Product Details

SNP ID
rs189874834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:129908320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGATGTGGAAGTAGGAAGATGCG[C/T]TCGCCGTTTTGGTGGGACTCAGTGT
Phenotype
MIM: 609963
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHSY3 PubMed Links

Gene Details

Gene
CHSY3
Gene Name
chondroitin sulfate synthase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175856.4 1420 Missense Mutation GCT,GTT A349V NP_787052.3
XM_005271982.3 1420 Missense Mutation GCT,GTT A349V XP_005272039.1
XM_005271983.3 1420 Missense Mutation GCT,GTT A349V XP_005272040.1
XM_011543363.2 1420 Missense Mutation GCT,GTT A40V XP_011541665.1
XM_011543364.2 1420 Intron XP_011541666.1
XM_011543365.2 1420 Missense Mutation GCT,GTT A349V XP_011541667.1
XM_017009434.1 1420 Intron XP_016864923.1
XM_017009435.1 1420 Missense Mutation GCT,GTT A349V XP_016864924.1

View Full Product Details