Product Details

SNP ID
rs189664077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85696006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCTGGGCCGTGACTACAGGACC[G/T]GTCTGACGATAGTCCAAAAACTGAA
Phenotype
MIM: 188855
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GNLY PubMed Links

Gene Details

Gene
GNLY
Gene Name
granulysin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302758.1 347 Missense Mutation GGT,TGT G96C NP_001289687.1
NM_006433.4 347 Missense Mutation GGT,TGT G69C NP_006424.2
NM_012483.3 347 Missense Mutation GGT,TGT G54C NP_036615.2
XM_005264084.2 347 Missense Mutation GGT,TGT G96C XP_005264141.1
XM_005264085.2 347 Missense Mutation GGT,TGT G69C XP_005264142.1
XM_005264087.2 347 Missense Mutation GGT,TGT G54C XP_005264144.1

View Full Product Details