Product Details
- SNP ID
-
rs189703933
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:34957486 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATCAGAACATGGAAGACTAAAGG[C/T]GGCTTTATCATAAGGCCATTATCAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF792
PubMed Links
Gene Details
- Gene
- ZNF792
- Gene Name
- zinc finger protein 792
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175872.4 |
2756 |
UTR 3 |
|
|
NP_787068.3 |
View Full Product Details